Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alix Antibody (OTI1A4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP271537FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271537FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271537FR Supplier Novus Biologicals Supplier No. NBP271537FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alix Monoclonal specifically detects Alix in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Alix
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A4
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein
Gene Symbols PDCD6IP
Host Species Mouse
Immunogen Full length recombinant protein of human Alix (NP_037506) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10015.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.