Learn More
Description
Specifications
Specifications
| Antigen | ALK-1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 24 |
| Conjugate | DyLight 680 |
| Dilution | ELISA, Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human ALK-1 (NP_000011.2; Met1-Gln118). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
