Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK-1 Antibody (124), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB088223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088223 Supplier Novus Biologicals Supplier No. NBP289257AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ALK-1
Applications Western Blot
Classification Monoclonal
Clone 124
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ALK-1 (NP_000011.2; Met1-Gln118).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 94
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.