Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK-7/Activin Receptor Type 1C Antibody (118311) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB377592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377592 Supplier Novus Biologicals Supplier No. FAB5771E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALK-7/Activin Receptor Type 1C Monoclonal antibody specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen ALK-7/Activin Receptor Type 1C
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 118311
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, ACTR-IC, ACVRLK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat ALK-7, Leu26-Arg108, Accession # NP_620790
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 130399
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.