Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK-7/Activin Receptor Type 1C Antibody (810506), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB311138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311138 Supplier Novus Biologicals Supplier No. FAB77491PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ALK-7/Activin Receptor Type 1C
Applications Flow Cytometry
Classification Monoclonal
Clone 810506
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, ACTR-IC, ACVRLK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ALK-7, Leu26-Glu113, Accession # Q8NER5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 130399
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.