Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK/CD246 Antibody (ALK/1032), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB384537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384537 Supplier Novus Biologicals Supplier No. NBP308772AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Western Blot

Specifications

Antigen ALK/CD246
Applications Western Blot
Classification Monoclonal
Clone ALK/1032
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Host Species Mouse
Immunogen Recombinant fragment (around aa 220-335) of human ALK/CD246 protein (exact sequence is proprietary) (Uniprot: Q9UM73)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 238
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.