Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALK, Mouse anti-Human, Clone: ALK1, Millipore Sigma™
Mouse Monoclonal Antibody
$192.34 - $498.79
Specifications
Antigen | ALK |
---|---|
Clone | ALK1 |
Dilution | Western Blotting Analysis: A representative lot detected ALK in Western Blotting applications (Pulford, K., et. al. (2002). Br J Haematol. 116(1):135-41; Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunohistochemistry Analysis: A representative lot detected ALK in Immunohistochemistry applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunofluorescence Analysis: A representative lot detected ALK in Immunofluorescence applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunoprecipitation Analysis: A representative lot immunoprecipitated ALK in Immunoprecipitation applications (Pulford, K., et. al. (2002). Br J Haematol. 116(1):135-41; Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunohistochemistry (Paraffin) Analysis: A representative lot detected ALK in Immunohistochemistry applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404). |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC117125
|
MilliporeSigma
MABC117125UG |
25 μg |
Each for $192.34
|
|
|||||
MABC1171100
|
MilliporeSigma
MABC1171100UG |
100 μg |
Each for $498.79
|
|
|||||
Description
Anti-ALK, clone ALK1, Cat. No. MABC1171, is a mouse monoclonal antibody that detects ALK tyrosine kinase receptor and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
ALK tyrosine kinase receptor (UniProt: Q9UM73; also known as EC: 2.7.10.1, Anaplastic lymphoma kinase, CD246) is encoded by the ALK gene (Gene ID: 238) in human. ALK tyrosine kinase receptor is a single-pass type I membrane protein that is essentially and transiently expressed in specific regions of the central and peripheral nervous systems and plays an important role in the development and function of the nervous system. It is predominantly expressed in the central nervous system, intestine, and testis. It acts as a receptor for ligands pleiotrophin (PTN) and midkine (MDK). It is activated and homodimerizes by ligand-binding and subsequent phosphorylation. Following activation by ligand, ALK induces tyrosine phosphorylation of CBL, FRS2, IRS1 and SHC1, as well as of the MAP kinases MAPK1/ERK2 and MAPK3/ERK1. It is inactivated through dephosphorylation by receptor protein tyrosine phosphatase beta and zeta complex (PTPRB/PTPRZ1) in the absence of any stimulation by a ligand. A chromosomal aberration involving ALK has been reported in non-Hodgkin lymphoma where the kinase is constitutively activated. Another chromosomal aberration involving ALK has also been linked with anaplastic large-cell lymphoma (ALCL). ALK tyrosine kinase receptor is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off in the mature form. ALK tyrosine kinase receptor consists of an extracellular domain (aa 19-1038); a transmembrane domain (aa 1039-1059); and a cytoplasmic domain (aa 1060-1620).Specifications
ALK | |
Western Blotting Analysis: A representative lot detected ALK in Western Blotting applications (Pulford, K., et. al. (2002). Br J Haematol. 116(1):135-41; Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunohistochemistry Analysis: A representative lot detected ALK in Immunohistochemistry applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunofluorescence Analysis: A representative lot detected ALK in Immunofluorescence applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunoprecipitation Analysis: A representative lot immunoprecipitated ALK in Immunoprecipitation applications (Pulford, K., et. al. (2002). Br J Haematol. 116(1):135-41; Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunohistochemistry (Paraffin) Analysis: A representative lot detected ALK in Immunohistochemistry applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404). | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ALK tyrosine kinase receptor;EC: 2.7.10.1;Anaplastic lymphoma kinase;CD246 | |
ALK | |
IgG3 κ | |
Protein G purified | |
Clone ALK1 specifically detects human ALK tyrosine kinase receptor. It targets an epitope with in 102 amino acids from the C-terminal region. |
ALK1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Signaling | |
Purified mouse monoclonal antibody IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_004295 | |
His-tagged full length human recombinant ALK tyrosine kinase receptor. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title