Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase/ALPL Antibody (B4-78) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB399963 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399963 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399963 Supplier Novus Biologicals Supplier No. FAB1448K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alkaline Phosphatase/ALPL Monoclonal antibody specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF

Specifications

Antigen Alkaline Phosphatase/ALPL
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone B4-78
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935
Host Species Mouse
Immunogen Human liver, bone and kidney-derived Alkaline Phosphatase/ALPL
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 249
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.