Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase/ALPP Antibody (8B6), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N103638P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N103638P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N103638P594 Supplier Novus Biologicals Supplier No. NB1103638PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Alkaline Phosphatase/ALPP
Applications Flow Cytometry
Classification Monoclonal
Clone 8B6
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme
Gene Symbols ALPP
Host Species Mouse
Immunogen Hep-2 cells with boosted surface expression of Alkaline Phosphatase, Placental. [UniProt No. P05187].
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 250.0
Test Specificity Alkaline Phosphatase, Placental - both Regan and Nagao isoenzymes. No cross reactivity with other isoenzymes of Alkaline Phosphatase.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.