Learn More
Alkaline Phosphatase/ALPP Antibody (ALPP/238), Alexa Fluor™ 750, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence |
| Classification | Monoclonal |
| Clone | ALPP/238 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme |
| Gene Symbols | ALPP |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.