Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase/ALPP Antibody (H17E2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB600750646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600750646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600750646 Supplier Novus Biologicals Supplier No. NB600750JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen AlkalinePhosphatase/ALPP
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone H17E2
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme
Gene Symbols ALPP
Host Species Mouse
Immunogen Placental membrane (Human).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 250
Test Specificity This recognises human placental alkaline phosphate. The has been widely used in immunolocalisation studies of germ cell tumours and may also be used in ELISA based assays for the measurement of soluble PLAP.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.