Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase, Bacterial Antibody (L2), Novus Biologicals™
SDP

Catalog No. NBP229868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229868 0.1 mL
NB019494 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229868 Supplier Novus Biologicals Supplier No. NBP229868

Mouse Monoclonal Antibody

Alkaline Phosphatase, Bacterial Monoclonal antibody specifically detects Alkaline Phosphatase, Bacterial in E. coli samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Alkaline Phosphatase, Bacterial
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone L2
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Formulation Ascites with No Preservative
Gene Accession No. P00634
Gene Alias BAP, ECK0378, JW0374, Pho A, PhoA
Gene Symbols phoA
Host Species Mouse
Immunogen F201C1B.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 945041
Test Specificity Alkaline phosphatase (AP). MAB1012 has a high affinity and recognizes an AP determinant resistant to denaturation by SDS-PAGE. It is therefore ideally suited for sensitive and specific detection of AP fusion proteins by Western Blot analysis of E. coli transformants expressing fusion products.
Target Species E. coli
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.