Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase, Intestinal Antibody (001), Biotin, Novus Biologicals™
SDP

Catalog No. NB070856 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070856 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070856 Supplier Novus Biologicals Supplier No. NBP290196B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Alkaline Phosphatase, Intestinal Monoclonal specifically detects Alkaline Phosphatase, Intestinal in Human samples. It is validated for ELISA.

Specifications

Antigen Alkaline Phosphatase, Intestinal
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Biotin
Dilution ELISA
Formulation PBS
Gene Alias alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Alkaline Phosphatase, Intestinal (P09923; Met1-Asp503).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Ovarian Carcinoma Cell Markers, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 248
Test Specificity No cross-reactivity in ELISA with: Human ALPL
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.