Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase, Intestinal Antibody (AAP1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB1002638IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002638IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002638IR Supplier Novus Biologicals Supplier No. NB1002638IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alkaline Phosphatase, Intestinal Monoclonal specifically detects Alkaline Phosphatase, Intestinal in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Alkaline Phosphatase, Intestinal
Applications ELISA, Western Blot
Classification Monoclonal
Clone AAP1
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme
Gene Symbols ALPI
Host Species Mouse
Immunogen D98/AH-2 (HeLa) cells, which produce human intestinal alkaline phosphatase ectopically
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Embryonic Stem Cell Markers, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 248
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.