Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Liver/Bone/Kidney Antibody (BGN/03/662) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$170.97 - $492.89
Specifications
| Antigen | Alkaline Phosphatase, Liver/Bone/Kidney |
|---|---|
| Clone | BGN/03/662 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017766
![]() |
Novus Biologicals
NB100663850.025MG |
0.025 mg |
Each for $170.97
|
|
|||||
NB10066385
![]() |
Novus Biologicals
NB10066385 |
0.1 mg |
Each for $492.89
|
|
|||||
Description
Alkaline Phosphatase, Liver/Bone/Kidney Monoclonal specifically detects Alkaline Phosphatase, Liver/Bone/Kidney in Human, Bacteria samples. It is validated for Western Blot, ELISA.Specifications
| Alkaline Phosphatase, Liver/Bone/Kidney | |
| 1.0 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Bacteria | |
| ALPL, BALP | |
| ALPL | |
| IgG1 | |
| Protein A purified | |
| NB100-66385 recognizes bone specific alkaline phosphatase. |
| BGN/03/662 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P05186 | |
| 249 | |
| Alkaline phosphatase from human bone. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title