Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase, Liver/Bone/Kidney Antibody (BGN/03/662), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10066385R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10066385R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10066385R Supplier Novus Biologicals Supplier No. NB10066385R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alkaline Phosphatase, Liver/Bone/Kidney Monoclonal specifically detects Alkaline Phosphatase, Liver/Bone/Kidney in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Alkaline Phosphatase, Liver/Bone/Kidney
Applications ELISA, Western Blot
Classification Monoclonal
Clone BGN/03/662
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALPL, BALP
Gene Symbols ALPL
Host Species Mouse
Immunogen Alkaline phosphatase from human bone.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 249
Test Specificity NB100-66385 recognizes bone specific alkaline phosphatase.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.