Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALKBH4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP214737 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214737 0.1 mL
NB437737 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP214737 Supplier Novus Biologicals Supplier No. NBP214737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

ALKBH4 Polyclonal specifically detects ALKBH4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ALKBH4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9NXW9
Gene Alias ABH4, ALKBH4 alkB, alkylation repair homolog 4 (E. coli)
Gene Symbols ALKBH4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: TYRFIYCSDTGWAVGTEESDFEGWAFPFPGVMLIEDFVTREEEAELVRLMDRDPWKLSQSGRRKQDYGPKVN
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54784
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.