Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALKBH8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
Antigen | ALKBH8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18047320
![]() |
Novus Biologicals
NBP18047320UL |
20 μL | N/A | N/A | N/A | ||||
NBP180473
![]() |
Novus Biologicals
NBP180473 |
100 μL | N/A | N/A | N/A | ||||
Description
ALKBH8 Polyclonal specifically detects ALKBH8 in Human samples. It is validated for Western Blot.Specifications
ALKBH8 | |
Polyclonal | |
Rabbit | |
EAW67088 | |
91801 | |
Synthetic peptide directed towards the N terminal of human ALKBH8. Peptide sequence MDSNHQSNYKLSKTEKKFLRKQIKAKHTLLRHEGIETVSYATQSLVVANG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ABH8MGC10235, AlkB homologue 8, alkB, alkylation repair homolog 8 (E. coli), alkylated DNA repair protein alkB homolog 8, EC 1.14.11.-, EC 2.1.1.-, FLJ38204, Probable alpha-ketoglutarate-dependent dioxygenase ABH8, S-adenosyl-L-methionine-dependent tRNA methyltransferase ABH8 | |
ALKBH8 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title