Learn More
Allegro™ Ambient Air Driven Pump A‐4000AD, 4-Worker Connection
Portable air source for up to four full or half-face respirator users, or three hood users. Ideal for use when electric power is unavailable or undesirable.
$3796.00
Specifications
Material Metal base and housing, steel plumbing
Weight (English) 83 lb.
Product Type Low Pressure Air Pump
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-087453
missing translation for 'viewDocuments'
Allegro™
9850H
Each for $3,796.00
Only null left
Add to Cart
19-087454
missing translation for 'viewDocuments'
Allegro™
9850S
Each for $3,796.00
Only null left
Add to Cart
Description
Description
  • For four respirator users or three hood users
  • Rigid metal base and housing
  • Brass and galvanized steel plumbing
  • Choose from hansen or schrader fittings
  • Adjustable brass pressure relief valve
  • Pressure gauge 0-15 psi (0-1.03 bar)
  • Oil-less, rotary vane, direct driven pump
  • Inlet filter 50 Micron felt material with metal grid inlet filter housing
  • Exhaust filter 0.3 Micron HEPA
Specifications
Specifications
Metal base and housing, steel plumbing
83 lb.
Low Pressure Air Pump
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.