Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allergin-1 Antibody (767727) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB374331 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374331 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374331 Supplier Novus Biologicals Supplier No. FAB7638Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Allergin-1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 767727
Conjugate DyLight 680
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, Probable Mast Cell Antigen 32 Homolog
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Allergin-1, Met1-Lys227, Accession # Q7Z6M3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284021
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.