Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allergin-1 Mouse anti-Mouse, Unconjugated, Clone: TX83, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Allergin-1 |
---|---|
Clone | TX83 |
Applications | Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF23322MI
|
MilliporeSigma
MABF233225UL |
25 μL |
Each for $182.31
|
|
|||||
MABF23321MI
|
MilliporeSigma
MABF2332100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Mouse samples. It is validated for Flow Cytometry, Function Assay.Specifications
Allergin-1 | |
Flow Cytometry, Functional Assay | |
Unconjugated | |
Mouse | |
Mouse | |
Allergy inhibitory receptor 1;Mast cell antigen 32;MCA-32;Mast cell Ag-32;Mast cell immunoglobulin-like receptor 1 | |
BW5147 cells expressing Flag-tagged Allergin-1. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
27.37 kDa calculated. |
TX83 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_001028607 | |
Mirr1, Gm885, Mca32 | |
IgG1 κ | |
Protein G Purified | |
Clone TX83 is a mouse monoclonal antibody that detects murine Allergin-1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title