Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Ether 98.0+%, TCI America™
SDP

Catalog No. A0229100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0229100ML 100 mL
A022925ML 25 mL
A0229500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0229100ML Supplier TCI America Supplier No. A0229100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 557-40-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008650
InChI Key ATVJXMYDOSMEPO-UHFFFAOYSA-N
Synonym Diallyl Ether
PubChem CID 11194
IUPAC Name 3-prop-2-enoxyprop-1-ene
SMILES C=CCOCC=C

Specifications

Color Colorless
Boiling Point 94°C
Quantity 100 mL
UN Number 2360
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Ether
EINECSNumber (2)-2422
RTECSNumber KN7525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.