Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl glycidyl ether, 99+%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
106-92-3
C6H10O2
114.14
MFCD00005143
LSWYGACWGAICNM-UHFFFAOYSA-N
allyl glycidyl ether,glycidyl allyl ether,allylglycidaether,oxirane, 2-propenyloxy methyl,allyl 2,3-epoxypropyl ether,allil-glicidil-etere,neoallyl g,1,2-epoxy-3-allyloxypropane,1-allyloxy-2,3-epoxypropane,santolin xi
7838
2-(prop-2-enoxymethyl)oxirane
C=CCOCC1CO1
106-92-3
C6H10O2
114.14
MFCD00005143
LSWYGACWGAICNM-UHFFFAOYSA-N
allyl glycidyl ether,glycidyl allyl ether,allylglycidaether,oxirane, 2-propenyloxy methyl,allyl 2,3-epoxypropyl ether,allil-glicidil-etere,neoallyl g,1,2-epoxy-3-allyloxypropane,1-allyloxy-2,3-epoxypropane,santolin xi
7838
2-(prop-2-enoxymethyl)oxirane
C=CCOCC1CO1
Specific Gravity 0.962
Boiling Point 154.0°C
Melting Point -100.0°C
Flash Point 48°C
Color Colorless
Packaging AcroSeal™ Glass Bottle
Quantity 100mL
Refractive Index 1.4332 to 1.4352
Water 0.2% Max. (K.F.)
Infrared Spectrum Authentic
Density 0.9620g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether and,benzene
Formula Weight 114.14
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Allyl glycidyl ether
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irri

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 203-442-4