Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl methyl carbonate, 98%

Catalog No. AC389750050
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle

Catalog No. AC389750050

Supplier: Thermo Scientific Chemicals 389750050

Only null left
Add to Cart

CAS: 35466-83-2 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35466-83-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00134545
InChI Key YHLVIDQQTOMBGN-UHFFFAOYSA-N
Synonym allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa
PubChem CID 534788
IUPAC Name methyl prop-2-enyl carbonate
SMILES COC(=O)OCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0220g/mL
Boiling Point 131.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Linear Formula CH2=CHCH2OCO2CH3
Quantity 5 mL
Specific Gravity 1.022
Formula Weight 116.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl methyl carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.