Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl phenyl sulfone, 98%

Catalog No. AAA1040114
Click to view available options
Quantity:
5 g
25 g

CAS: 16212-05-8 | C9H10O2S | 182.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16212-05-8
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00014739
InChI Key KYPIULIVYSQNNT-UHFFFAOYSA-N
Synonym allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
PubChem CID 85330
IUPAC Name prop-2-enylsulfonylbenzene
SMILES C=CCS(=O)(=O)C1=CC=CC=C1

Specifications

Density 1.189
Boiling Point 111°C to 113°C (0.5 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1863561
Sensitivity Light sensitive
Formula Weight 182.24
Percent Purity 98%
Chemical Name or Material Allyl phenyl sulfone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-338-8
RTECSNumber WR2400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.