Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylurea 95%, Thermo Scientific™

Catalog No. AC103050250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103050250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103050250 Supplier Thermo Scientific Chemicals Supplier No. 103050250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-11-9
Molecular Formula C4H8N2O
Molecular Weight (g/mol) 100.121
MDL Number MFCD00007954
InChI Key VPJDULFXCAQHRC-UHFFFAOYSA-N
Synonym allylurea, 1-allylurea, n-allylurea, urea, 2-propenyl, allylcarbamide, monoallylurea, allyl urea, n-2-propenylurea, urea, allyl, 2-propenylurea
PubChem CID 11181
IUPAC Name prop-2-enylurea
SMILES C=CCNC(=O)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2C=CHCH2NHCONH2
Quantity 25 g
Beilstein 04, 209
Merck Index 15, 291
Formula Weight 100.12
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Allylurea, 95%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.