Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Alnico Bar Magnets



These Alnico Bar Magnets are powerful magnets for classroom experiments.
$5.50 - $18.00
Specifications
Grade | 5+ |
---|---|
Product Type | Magnet |
For Use With (Application) | Magnetic Demonstrations |
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
S15705
![]() ![]() |
United Scientific™
ABM015 |
1.5 x 0.5 x .25 in. |
Pair for $5.50
|
|
|||||
S15704
![]() ![]() |
United Scientific™
ABM020 |
2 x 0.5 x .25 in. |
Pair for $7.90
|
|
|||||
S15862
![]() ![]() |
United Scientific™
ABM030 |
3 x 0.5 x .25 in. |
Pair for $9.65
|
|
|||||
S15773
![]() ![]() |
United Scientific™
ABM040 |
4 x 0.5 x .25 in. |
Pair for $18.00
|
|
|||||
Description
- Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers.
- Magnets are painted with North and South poles, clearly marked.
- Magnets available in five different sizes.
Specifications
5+ | |
Magnet | |
Magnetic Demonstrations |
Warning: Cancer and reproductive harm
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title