Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALOX15 Monoclonal Antibody (OTI8C5), TrueMAB™, OriGene
SDP

Catalog No. p-200013122
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0429 30 μL
50-167-4975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0429 Supplier OriGene Supplier No. TA504249S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALOX15 Monoclonal antibody specifically detects ALOX15 in Human samples. It is validated for Western Blot

Specifications

Antigen ALOX15
Applications Western Blot
Classification Monoclonal
Clone OTI8C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C1S
Gene Accession No. P09871
Gene Alias 12/15-lipoxygenase; 12/15-LO; 12/15-LOX; 12-LO; 12-LOX; 15-lipooxygenase-1; 15-LOX; 15-LOX-1; 15LOX-1; 15-LOX2; Alox12; Alox12l; Alox15; Arachidonate 12-lipoxygenase, leukocyte-type; arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; L-12LO; LOG15
Host Species Mouse
Immunogen Full length human recombinant protein of human C1S produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 716
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.