Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha 1 Mannosidase 1A Antibody, Novus Biologicals™
SDP

Catalog No. NB436367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436367 25 μL
NBP237938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436367 Supplier Novus Biologicals Supplier No. NBP23793825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

alpha 1 Mannosidase 1A Polyclonal specifically detects alpha 1 Mannosidase 1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen alpha 1 Mannosidase 1A
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P33908
Gene Alias Alpha-1,2-mannosidase IA, EC 3.2.1, EC 3.2.1.113, HUMM3, HUMM9, Man(9)-alpha-mannosidase, MAN9, Man9-mannosidase, Mannosidase alpha class 1A member 1, mannosidase, alpha, class 1A, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IA, Processing alpha-1,2-mannosidase IA
Gene Symbols MAN1A1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: TLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREK
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 4121
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.