Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha 1 Mannosidase 1A Antibody, Novus Biologicals™
SDP

Catalog No. NB600977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600977 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600977 Supplier Novus Biologicals Supplier No. NB600977

Rabbit Polyclonal Antibody

alpha 1 Mannosidase 1A Polyclonal specifically detects alpha 1 Mannosidase 1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha 1 Mannosidase 1A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-10 ug/ml
Formulation 10mM PBS (pH 7.4) and 1.0% BSA with 15mM Sodium Azide
Gene Accession No. P33908
Gene Alias Alpha-1,2-mannosidase IA, EC 3.2.1, EC 3.2.1.113, HUMM3, HUMM9, Man(9)-alpha-mannosidase, MAN9, Man9-mannosidase, Mannosidase alpha class 1A member 1, mannosidase, alpha, class 1A, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IA, Processing alpha-1,2-mannosidase IA
Gene Symbols MAN1A1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acides 138-154 of human alpha 1,2 Mannosidase IA, with N-terminal added cysteine, conjugated to KLH.
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 4121
Test Specificity Detects alpha 1,2-Mannosidase IA.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.