Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha 1-Microglobulin Sheep anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB017435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017435 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017435 Supplier Novus Biologicals Supplier No. NB100621650.1ML
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

alpha 1-Microglobulin Polyclonal specifically detects alpha 1-Microglobulin in Human samples. It is validated for ELISA, Immunodiffusion.

Specifications

Antigen alpha 1-Microglobulin
Applications ELISA, Immunodiffusion
Classification Polyclonal
Concentration 5.0 mg/ml
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunodiffusion
Formulation PBS
Gene Accession No. P02760
Gene Alias A1M, alpha-1-microglobulin/bikunin precursor, bikunin, EDC1, HCPcomplex-forming glycoprotein heterogeneous in charge, HI30, IATIL, inter-alpha-trypsin inhibitor light chain, ITI, ITILC, ITILgrowth-inhibiting protein 19, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein, UTI
Host Species Sheep
Immunogen Highly pure human alpha-1-microglobulin, prepared from the urine of patients with chronic renal proteinuria (molecular weight 25-30 kD).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.