Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1A Adrenergic R/ADRA1A Antibody (518508) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB401933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401933 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401933 Supplier Novus Biologicals Supplier No. FAB10298I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-1A Adrenergic R/ADRA1A Monoclonal antibody specifically detects alpha-1A Adrenergic R/ADRA1A in Human samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen alpha-1A Adrenergic R/ADRA1A
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 518508
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human alpha-1 Adrenergic R/ADRA1A, Accession # P35348
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 148
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.