Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1B Adrenergic R/ADRA1B Antibody (471802), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB369125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369125 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369125 Supplier Novus Biologicals Supplier No. FAB4730W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-1B Adrenergic R/ADRA1B Monoclonal specifically detects alpha-1B Adrenergic R/ADRA1B in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen alpha-1B Adrenergic R/ADRA1B
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 471802
Conjugate DyLight 650
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ADRA1, adrenergic, alpha-1B-, receptor, alpha-1B adrenergic receptor, Alpha-1B adrenoceptor, Alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor, ALPHA1BAR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human ADRA1B, Met1-Phe520, Accession # P35368
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 147
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.