Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1B Adrenergic R/ADRA1B Antibody - Azide and BSA Free
SDP

Catalog No. NB12015851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12015851 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12015851 Supplier Novus Biologicals Supplier No. NB12015851

Chicken Polyclonal Antibody

alpha-1B Adrenergic R/ADRA1B Polyclonal specifically detects alpha-1B Adrenergic R/ADRA1B in Human samples. It is validated for Western Blot, Flow (Cell Surface).

Specifications

Antigen alpha-1B Adrenergic R/ADRA1B
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow (Cell Surface)
Formulation PBS with No Preservative
Gene Alias ADRA1, adrenergic, alpha-1B-, receptor, alpha-1B adrenergic receptor, Alpha-1B adrenoceptor, Alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor, ALPHA1BAR
Gene Symbols ADRA1B
Host Species Chicken
Immunogen Synthetic peptide, corresponding to amino acids 200-299 of Human Alpha 1b Adrenergic Receptor.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 147
Test Specificity Alpha 1b Adrenergic Receptor
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.