Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha-2-HS-Glycoprotein, Rabbit anti-Human, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
$192.34 - $525.84
Specifications
Antigen | Alpha-2-HS-Glycoprotein |
---|---|
Dilution | Western Blotting Analysis: A 1:500 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human retina tissue lysate.Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human liver and human bone marrow tissue sections. |
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN146925UL
|
MilliporeSigma
ABN146925UL |
25 μL |
Each for $192.34
|
|
|||||
ABN1469100
|
MilliporeSigma
ABN1469100UL |
100 μL |
Each for $525.84
|
|
|||||
Description
Anti-Alpha-2-HS-Glycoprotein Antibody, Cat. No. ABN1469, is a highly specific rabbit polyclonal antibody that targets Alpha-2-HS-Glycoprotein and has been tested for use Immunohistochemistry and Western Blotting.
Alpha-2-HS-glycoprotein (UniProt: P02765; also known as Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, Fetuin-A, Alpha-2 Heremans-Schmid glycoprotein, AHSG) is encoded by the AHSG (also known as FETUA, PRO2743) gene (Gene ID: 197) in human. Alpha-2-HS-glycoprotein is a serum protein of the cystatin superfamily that is synthesized by hepatocytes and is considered as a negative acute-phase reactant. It is highly expressed in various fetal tissues, but in adults it is mainly expressed in the liver. It is synthesized with a signal peptide (aa 1-18) and a connective propeptide (aa 301-340) and is subsequently cleaved into 2 chains known as Alpha-2-HS-glycoprotein chain A and Alpha-2-HS-glycoprotein chain B. The mature product is involved in multiple normal biological and pathological functions. It promotes possesses opsonic properties and is shown to be involved in endocytosis, brain development, bone resorption, regulation of insulin activity and hepatocyte growth factor (HGF) activity. Alpha-2-HS-glycoprotein also serves as a carrier protein and serves as a carrier for otherwise insoluble calcium phosphate and can act as a potent inhibitor of pathological vascular calcification. Higher levels of Alpha-2-HS-glycoprotein have been reported in gestational and type II diabetes. It is also present at higher levels in patients with endometriosis. Lower levels of Alpha-2-HS-glycoprotein are present in cancer subjects. (Ref.: Kalabay, L., et al. (2002). Eur. J. Endocrinol. 147(2); 243-248; Sindhu, S., et al. (2016). BMC Immunol. 17; 33).Specifications
Alpha-2-HS-Glycoprotein | |
Immunohistochemistry, Western Blot | |
Unconjugated | |
RUO | |
Human | |
Alpha-2-Z-globulin;Ba-alpha-2-glycoprotein;Fetuin-A;Alpha-2 Heremans-Schmid glycoprotein;AHGS | |
AHSG;FETUA;PRO2743 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
∽55 kDa observed; 39.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Western Blotting Analysis: A 1:500 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human retina tissue lysate.Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human liver and human bone marrow tissue sections. | |
Polyclonal | |
Rabbit | |
Neuroscience | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_001613.2 | |
GST/His-tagged recombinant fragment corresponding to 146 amino acids from human Alpha-2-HS-glycoprotein. | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects human Alpha-2-HS-glycoprotein. It targets an epitope with in the N-terminal half. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title