Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha 2-Macroglobulin Antibody (A2M/3623), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB391383 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391383 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391383 Supplier Novus Biologicals Supplier No. NBP308689V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen alpha 2-Macroglobulin
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone A2M/3623
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7
Host Species Mouse
Immunogen Human recombinant alpha 2-Macroglobulin protein fragment (around aa 604-748) (exact sequence is proprietary) (Uniprot: P01023)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.