Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2A Adrenergic R/ADRA2A Antibody, Novus Biologicals™
SDP

Catalog No. NBP222452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222452 Supplier Novus Biologicals Supplier No. NBP222452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

alpha-2A Adrenergic R/ADRA2A Polyclonal specifically detects alpha-2A Adrenergic R/ADRA2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen alpha-2A Adrenergic R/ADRA2A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000
Formulation PBS and 1 mg/ml BSA. with 0.05% Sodium Azide
Gene Alias ADRA2, ADRA2Ralpha-2-adrenergic receptor, platelet type, ADRARZNF32, adrenergic, alpha-2A-, receptor, Alpha-2 adrenergic receptor subtype C10, alpha-2A adrenergic receptor, Alpha-2A adrenoceptor, Alpha-2A adrenoreceptor, Alpha-2AAR, ALPHA2AAR, alpha-2AAR subtype C10
Gene Symbols ADRA2A
Host Species Rabbit
Immunogen Synthetic peptide corressponding to residues R(218) I Y Q I A K R R T R V P P S R R G(235) of the 3rd intracellular loop of human A2AAR.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.