Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2B Adrenergic R/ADRA2B Antibody (491613), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB350196 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350196 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350196 Supplier Novus Biologicals Supplier No. FAB10324MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-2B Adrenergic R/ADRA2B Monoclonal specifically detects alpha-2B Adrenergic R/ADRA2B in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen alpha-2B Adrenergic R/ADRA2B
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 491613
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ADRA2B adrenergic, alpha-2B-, receptor, ADRA2L1adrenergic receptor alpha 2B, ADRA2RL1, ADRARL1, adrenergic, alpha-2B-, receptor, Alpha-2 adrenergic receptor subtype C2, alpha-2-adrenergic receptor-like 1, alpha-2B adrenergic receptor, Alpha-2B adrenoceptor, Alpha-2B adrenoreceptor, alpha-2B-adrenergic receptor, Alpha-2BAR, ALPHA2BAR, G-protein coupled receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human alpha-2B Adrenergic R/ADRA2B, Accession # P18089
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 151
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.