Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2B Adrenergic R/ADRA2B Antibody (5G10), Novus Biologicals™
SDP

Catalog No. NBP266606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266606 Supplier Novus Biologicals Supplier No. NBP266606
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-2B Adrenergic R/ADRA2B Monoclonal specifically detects alpha-2B Adrenergic R/ADRA2B in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen alpha-2BAdrenergicR/ADRA2B
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5G10
Conjugate Unconjugated
Dilution Western Blot 5-10 ug/ml, Immunohistochemistry
Formulation PBS with 0.08% Sodium Azide
Gene Accession No. P19328
Gene Alias ADRA2B adrenergic, alpha-2B-, receptor, ADRA2L1adrenergic receptor alpha 2B, ADRA2RL1, ADRARL1, adrenergic, alpha-2B-, receptor, Alpha-2 adrenergic receptor subtype C2, alpha-2-adrenergic receptor-like 1, alpha-2B adrenergic receptor, Alpha-2B adrenoceptor, Alpha-2B adrenoreceptor, alpha-2B-adrenergic receptor, Alpha-2BAR, ALPHA2BAR, G-protein coupled receptor
Gene Symbols ADRA2B
Host Species Mouse
Immunogen Mouse monoclonal antibody raised to a synthetic peptide corresponding to the third intracellular loop of the rat alpha-2B Adrenergic R/ADRA2B (Uniprot: P19328).
Purification Method Protein A or G purified
Quantity 100 μg
Research Discipline Adaptive Immunity, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 151
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.