Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2C Adrenergic R/ADRA2C Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB345042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345042 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345042 Supplier Novus Biologicals Supplier No. NB10093554PCP

Rabbit Polyclonal Antibody

alpha-2C Adrenergic R/ADRA2C Polyclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen alpha-2C Adrenergic R/ADRA2C
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR
Host Species Rabbit
Immunogen Synthetic peptide comprising internal residues of the human Alpha 2c protein. Reacts with rat and mouse Alpha 2c.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 152
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.