Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Actinin 1 Antibody (OTI7A4), HRP, Novus Biologicals™
SDP

Catalog No. NBP270078H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270078H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270078H Supplier Novus Biologicals Supplier No. NBP270078H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Actinin 1 Monoclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen alpha-Actinin 1
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone OTI7A4
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Knockout Validated
Formulation PBS with No Preservative
Gene Alias actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1
Gene Symbols ACTN1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 650-893 of human alpha-Actinin 1 (NP_001093) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 87.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.