Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Actinin 1 Mouse anti-Human, Mouse, Clone: 1D10, Novus Biologicals™
SDP

Catalog No. NBP25946550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25946550 50 μL
NBP259465X 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25946550 Supplier Novus Biologicals Supplier No. NBP25946550UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-Actinin 1 Monoclonal specifically detects alpha-Actinin 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha-Actinin 1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1065, ELISA
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1
Gene Symbols ACTN1
Host Species Mouse
Immunogen Recombinant human ACTN1 (1-249aa) purified from E. coli
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 87
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.