Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Actinin 2 Antibody (EA-53), Novus Biologicals™
SDP

Catalog No. NBP122630 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122630 0.5 mL
NB018463 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP122630 Supplier Novus Biologicals Supplier No. NBP122630
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Alpha Actinin 2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EA-53
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20-1:30, Immunohistochemistry-Paraffin 1:20-1:30
Formulation Ascites with 0.05% Sodium Azide
Gene Alias actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein
Gene Symbols ACTN2
Host Species Mouse
Immunogen Purified rabbit skeletal alpha-actinin.
Purification Method Ascites
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 88
Test Specificity This antibody is specific for alpha-skeletal muscle and alpha-cardiac actinin. It stains Z lines and dots in stress fibers of myotubes in skeletal and cardiac muscle but not in non-sarcomeric muscle elements. This antibody shows a wide range of cross reactivity with various mammalian species.
Target Species Human
Content And Storage Store at 4°C.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.