Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 2 Antibody (EA-53), Novus Biologicals™

Mouse Monoclonal Antibody
$192.94 - $656.50
Specifications
| Antigen | Alpha Actinin 2 |
|---|---|
| Clone | EA-53 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Alpha Actinin 2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| 88 | |
| Purified rabbit skeletal alpha-actinin. | |
| Primary | |
| Store at 4°C. |
| EA-53 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
| ACTN2 | |
| IgG1 | |
| Ascites | |
| This antibody is specific for alpha-skeletal muscle and alpha-cardiac actinin. It stains Z lines and dots in stress fibers of myotubes in skeletal and cardiac muscle but not in non-sarcomeric muscle elements. This antibody shows a wide range of cross reactivity with various mammalian species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title