Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Actinin 4 Antibody (93), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB387152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387152 Supplier Novus Biologicals Supplier No. NBP308415R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alpha Actinin 4 Monoclonal antibody specifically detects Alpha Actinin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Alpha Actinin 4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 93
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4
Host Species Mouse
Immunogen Recombinant full-length human Alpha Actinin 4 protein (Uniprot: O43707)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.