Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Adaptin Antibody (AP6), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB6001545G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001545G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001545G Supplier Novus Biologicals Supplier No. NB6001545G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha Adaptin Monoclonal specifically detects alpha Adaptin in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha Adaptin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AP6
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Alias 100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit
Gene Symbols AP2A1
Host Species Mouse
Immunogen Purified adaptors.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160
Test Specificity Detects assembly polypeptide 2 (AP2) It recognizes the products of both alpha-adaptin genes, alpha A and alpha C as well as an alternatively spliced isoform of alpha A found in neurons.
Target Species Human, Mouse, Rat, Bovine, Hamster, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.