Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Aminoadipate Aminotransferase Antibody, Novus Biologicals™
SDP

Catalog No. NB11099587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11099587 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11099587 Supplier Novus Biologicals Supplier No. NB11099587
Add to Cart
Add to Cart

Goat Polyclonal Antibody

alpha-Aminoadipate Aminotransferase Polyclonal specifically detects alpha-Aminoadipate Aminotransferase in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen alpha-Aminoadipate Aminotransferase
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_057312.1
Gene Alias AadAT, aminoadipate aminotransferase, EC 2.6.1.39,2-aminoadipate aminotransferase, EC 2.6.1.7,2-aminoadipate transaminase, KAT/AadAT, KAT2mitochondrial, Kynurenine aminotransferase II, Kynurenine--oxoglutarate aminotransferase II, Kynurenine--oxoglutarate transaminase II, L kynurenine/alpha aminoadipate aminotransferase
Gene Symbols AADAT
Host Species Goat
Immunogen Peptide with sequence C-KPEDAKNPQKNTPK corresponding to internal region according to NP_057312.1, NP_872603.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51166
Test Specificity Both reported variants represent identical protein (NP_057312.1and NP_872603.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.