Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Catenin Monoclonal Antibody (alpha-CAT-7A4)
GREENER_CHOICE

Catalog No. 139700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-970-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-970-0 Supplier Invitrogen™ Supplier No. 139700
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha Catenin Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC, IP, Western Blot

Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Catenin
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone alpha-CAT-7A4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CTNNA1
Gene Accession No. P26231, P26232, P30997, P35221, Q61301
Gene Alias 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI481747; AI988031; alpha E catenin; alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; alpha-catenin-related protein; alpha-E-catenin; alphaE-catenin; alpha-N-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; CAPR; CAP-R; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha1; catenin alpha-1; Catenin alpha2; Catenin alpha-2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; Catna; Catna1; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA1; Ctnna2; CTNR; FLJ36832; FLJ52416; HGNC:2509; MDPT2; Neural alpha-catenin; NY-REN-13 antigen; renal carcinoma antigen NY-REN-13
Gene Symbols CTNNA1, CTNNA2
Host Species Mouse
Immunogen Synthetic peptide derived from the C-terminus of mouse alpha-catenin.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12385, 12386, 1495, 1496, 297357, 307505, 396035, 416184
Target Species Human, Mouse, Rat, Chicken, Frog
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.