Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha COP I Antibody, Novus Biologicals™
SDP

Catalog No. NB1202913 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202913 Supplier Novus Biologicals Supplier No. NB1202913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

alpha COP I Polyclonal specifically detects alpha COP I in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen alpha COP I
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 3 ug/ml
Formulation PBS, 1mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. G3V6T1
Gene Alias alpha coat protein, Alpha-coat protein, Alpha-COP, coatomer protein complex, subunit alpha, coatomer subunit alpha, FLJ26320, HEPCOP, HEP-COPalpha-COP, xenin
Gene Symbols COPA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) L T K F E T K S A R V K G L S F H P(19) C of rat alpha-COP.
Molecular Weight of Antigen 138 kDa
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1314
Test Specificity Detects coatomer-protein I alpha (COP I alpha).
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.