Learn More
Invitrogen™ alpha COP Recombinant Rabbit Monoclonal Antibody (3W6Y6)

Description
Positive Samples: HeLa, HT-29, MCF7, Mouse lung, Rat brain Immunogen sequence: MLTKFETKSA RVKGLSFHPK RPWILTSLHN GVIQLWDYRM CTLIDKFDEH DGPVRGIDFH KQQPLFVSGG DDYKIKVWNY KLRRCLFTLL GHLDYIRTTF.
Specifications
Specifications
| Antigen | alpha COP |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 3W6Y6 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | COPA |
| Gene Accession No. | P53621, Q8CIE6 |
| Gene Alias | AILJK; alpha coat protein; alpha-coat protein; Alpha-COP; AU040324; coatomer protein complex subunit alpha; coatomer subunit alpha; COPA; HEPCOP; HEP-COP; proxenin; xenin; Xenopsin-related peptide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.